siRNA Details
| Gene Symbol: SPRR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003125.2 | NP_003116.2 | 2 | 558 | ||
| GenBank mRNA | BC056240.1 | - | 2 | 562 | ||
| M19888.1 | - | 2 | 562 | |||
| M21300.1 | - | 2 | 558 | |||
| M21301.1 | - | 2 | 558 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPRR1B| Gene Symbol: SPRR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003125.2 | NP_003116.2 | 2 | 558 | ||
| GenBank mRNA | BC056240.1 | - | 2 | 562 | ||
| M19888.1 | - | 2 | 562 | |||
| M21300.1 | - | 2 | 558 | |||
| M21301.1 | - | 2 | 558 | |||