siRNA Details
| Gene Symbol: Arl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022385.3 | NP_071780.1 | 4 | 334 | ||
| GenBank mRNA | BC061553.1 | - | 417 | 6889396 | ||
| FQ226660.1 | - | 4 | 390 | |||
| U12402.1 | - | 4 | 266 | |||
| X76920.1 | - | 4 | 334 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arl1| Gene Symbol: Arl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022385.3 | NP_071780.1 | 4 | 334 | ||
| GenBank mRNA | BC061553.1 | - | 417 | 6889396 | ||
| FQ226660.1 | - | 4 | 390 | |||
| U12402.1 | - | 4 | 266 | |||
| X76920.1 | - | 4 | 334 | |||