siRNA Details
| Gene Symbol: Arl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022385.3 | NP_071780.1 | 2, 3 | 244 | ||
| GenBank mRNA | BC061553.1 | - | 327 | 6889396 | ||
| FQ226660.1 | - | 2, 3 | 300 | |||
| U12402.1 | - | 2, 3 | 176 | |||
| X76920.1 | - | 2, 3 | 244 | |||

Arl1
Genomic Map