siRNA Details
| Gene Symbol: Mafg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022386.2 | NP_071781.1 | 3 | 1583 | ||
| XM_006247912.2 | XP_006247974.1 | 3 | 5666 | |||
| XM_006247913.2 | XP_006247975.1 | 4 | 4667 | |||
| XM_006247914.2 | XP_006247976.1 | 2 | 1586 | |||
| XM_006247916.2 | XP_006247978.1 | 2 | 3363 | |||
| XM_006247917.2 | XP_006247979.1 | 2 | 6135 | |||
| XM_006247918.2 | XP_006247980.1 | 2 | 2746 | |||
| XM_006247919.2 | XP_006247981.1 | 2 | 3293 | |||
| XM_006247921.2 | XP_006247983.1 | 3 | 2344 | |||
| XM_006247922.2 | XP_006247984.1 | 3 | 1505 | |||
| XM_006247923.2 | XP_006247985.1 | 3 | 2274 | |||
| XM_008768482.1 | XP_008766704.1 | 3 | 5116 | |||
| GenBank mRNA | BC078828.1 | - | 3 | 1583 | 7127046 | |

Mafg
Genomic Map