siRNA Details
| Gene Symbol: Pafah1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022387.3 | NP_071782.1 | 5 | 440 | ||
| XM_006242957.2 | XP_006243019.1 | 5 | 906 | |||
| XM_006242958.2 | XP_006243020.1 | 5 | 439 | |||
| XM_008766172.1 | XP_008764394.1 | 5 | 670 | |||
| GenBank mRNA | AF016048.1 | - | 5 | 403 | ||
| BC081714.1 | - | 5 | 440 | 7108923 | ||
| FQ212439.1 | - | 5 | 727 | |||

Pafah1b2
Genomic Map