siRNA Details
| Gene Symbol: Pttg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022391.2 | NP_071786.1 | 3 | 561 | ||
| XM_006246142.1 | XP_006246204.1 | 4 | 372 | |||
| XM_006246143.2 | XP_006246205.1 | 4 | 569 | |||
| XM_006246144.1 | XP_006246206.1 | 4 | 666 | |||
| XM_006246145.2 | XP_006246207.1 | 4 | 372 | |||
| XM_008767643.1 | XP_008765865.1 | 4 | 321 | |||
| GenBank mRNA | BC158551.1 | - | 4 | 671 | 9032805 | |
| U73030.1 | - | 3 | 573 | |||

Pttg1
Genomic Map