siRNA Details
| Gene Symbol: Insig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022392.1 | NP_071787.1 | 4 | 781 | ||
| GenBank mRNA | BC078827.1 | - | 4 | 791 | 7125208 | |
| L13619.1 | - | 4 | 781 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Insig1| Gene Symbol: Insig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022392.1 | NP_071787.1 | 4 | 781 | ||
| GenBank mRNA | BC078827.1 | - | 4 | 791 | 7125208 | |
| L13619.1 | - | 4 | 781 | |||