siRNA Details
| Gene Symbol: Gng11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022396.2 | NP_071791.1 | 2 | 420 | ||
| GenBank mRNA | AF257110.1 | - | 2 | 141 | ||
| BC097320.1 | - | 2 | 420 | 7455035 | ||
| Gene Symbol: LOC100912034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749698.3 | XP_003749746.1 | 2 | 578 | ||

Gng11
Genomic Map