siRNA Details
| Gene Symbol: Hnrnpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037285.1 | NP_001032362.1 | 4 | 1383 | ||
| NM_001037286.1 | NP_001032363.1 | 3 | 1425 | |||
| NM_001037287.1 | NP_001032364.1 | 4 | 1340 | |||
| NM_022397.3 | NP_071792.1 | 4 | 1354 | |||
| XM_008763262.1 | XP_008761484.1 | 1 | 1112 | |||
| GenBank mRNA | AB022209.1 | - | 1 | 1077 | ||
| BC097275.1 | - | 4 | 1323 | 7444983 | ||
| BC103634.1 | - | 4 | 1342 | 7097633 | ||

Hnrnpf
Genomic Map