siRNA Details
| Gene Symbol: Slc25a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022398.2 | NP_071793.1 | 5, 6 | 647 | ||
| GenBank mRNA | FQ216657.1 | - | 675 | |||
| U84727.1 | - | 5, 6 | 647 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a11| Gene Symbol: Slc25a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022398.2 | NP_071793.1 | 5, 6 | 647 | ||
| GenBank mRNA | FQ216657.1 | - | 675 | |||
| U84727.1 | - | 5, 6 | 647 | |||