siRNA Details
| Gene Symbol: Calr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022399.2 | NP_071794.1 | 8 | 1042 | ||
| GenBank mRNA | BC062395.1 | - | 8 | 1019 | 5623178 | |
| D78308.1 | - | 8 | 994 | |||
| FQ210587.1 | - | 1047 | ||||
| FQ226996.1 | - | 2 | 127 | |||
| X13702.1 | - | 2 | 175 | |||
| X53363.1 | - | 8 | 1014 | |||
| X79327.1 | - | 8 | 992 | |||

Calr
Genomic Map