siRNA Details
| Gene Symbol: Calr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022399.2 | NP_071794.1 | 4, 5 | 548 | ||
| GenBank mRNA | BC062395.1 | - | 4, 5 | 525 | 5623178 | |
| BU671467.1 | - | 4, 5 | 548 | |||
| D78308.1 | - | 4, 5 | 500 | |||
| FQ210587.1 | - | 553 | ||||
| FQ220096.1 | - | 4, 5 | 554 | |||
| X53363.1 | - | 4, 5 | 520 | |||
| X79327.1 | - | 4, 5 | 498 | |||

Calr
Genomic Map