siRNA Details
| Gene Symbol: Bcat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022400.1 | NP_071795.1 | 7 | 818 | ||
| XM_006229149.1 | XP_006229211.1 | 7 | 995 | |||
| XM_008759413.1 | XP_008757635.1 | 7 | 1545 | |||
| GenBank mRNA | BC061790.1 | - | 7 | 799 | 5623666 | |
| U68417.1 | - | 7 | 818 | |||

Bcat2
Genomic Map