siRNA Details
| Gene Symbol: SPRR2F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014450.2 | NP_001014450.1 | 2 | 558 | ||
| XM_017002176.1 | XP_016857665.1 | 2 | 624 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPRR2F| Gene Symbol: SPRR2F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014450.2 | NP_001014450.1 | 2 | 558 | ||
| XM_017002176.1 | XP_016857665.1 | 2 | 624 | |||