siRNA Details
| Gene Symbol: Oat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022521.3 | NP_071966.1 | 7 | 938 | ||
| GenBank mRNA | BC061551.1 | - | 7 | 941 | 6890520 | |
| FQ210816.1 | - | 1 | 39 | |||
| M11842.1 | - | 6 | 881 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Oat| Gene Symbol: Oat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022521.3 | NP_071966.1 | 7 | 938 | ||
| GenBank mRNA | BC061551.1 | - | 7 | 941 | 6890520 | |
| FQ210816.1 | - | 1 | 39 | |||
| M11842.1 | - | 6 | 881 | |||