siRNA Details
| Gene Symbol: Casp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022522.2 | NP_071967.2 | 8 | 907 | ||
| XM_006236394.2 | XP_006236456.1 | 7 | 1588 | |||
| XM_008762918.1 | XP_008761140.1 | 7 | 1588 | |||
| GenBank mRNA | AF025671.1 | - | 7 | 700 | ||
| AF136231.1 | - | 8 | 905 | |||
| AF136232.1 | - | 8 | 905 | |||
| U34684.1 | - | 3 | 310 | |||
| U77933.1 | - | 8 | 907 | |||

Casp2
Genomic Map