siRNA Details
| Gene Symbol: SPRR2G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014291.3 | NP_001014313.1 | 2 | 580 | ||
| XM_017002177.1 | XP_016857666.1 | 4 | 957 | |||
| XM_017002178.1 | XP_016857667.1 | 2 | 503 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPRR2G| Gene Symbol: SPRR2G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014291.3 | NP_001014313.1 | 2 | 580 | ||
| XM_017002177.1 | XP_016857666.1 | 4 | 957 | |||
| XM_017002178.1 | XP_016857667.1 | 2 | 503 | |||