siRNA Details
| Gene Symbol: Sncg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031688.1 | NP_113876.1 | 5 | 535 | ||
| GenBank mRNA | X86789.1 | - | 5 | 535 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sncg| Gene Symbol: Sncg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031688.1 | NP_113876.1 | 5 | 535 | ||
| GenBank mRNA | X86789.1 | - | 5 | 535 | ||