siRNA Details
| Gene Symbol: Crybb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031690.1 | NP_113878.1 | 1, 2 | 123 | ||
| XM_006249572.1 | XP_006249634.1 | 2, 3 | 161 | |||
| GenBank mRNA | AF287304.1 | - | 1, 2 | 95 | ||
| X05899.1 | - | 1, 2 | 123 | |||

Crybb3
Genomic Map