siRNA Details
| Gene Symbol: Itgad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031691.1 | NP_113879.1 | 9, 10 | 1023 | ||
| GenBank mRNA | AF021334.1 | - | 9, 10 | 1023 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itgad| Gene Symbol: Itgad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031691.1 | NP_113879.1 | 9, 10 | 1023 | ||
| GenBank mRNA | AF021334.1 | - | 9, 10 | 1023 | ||