siRNA Details
Gene Symbol: Ykt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031692.2 | NP_113880.2 | 5 | 446 | ||
GenBank mRNA | AF033027.1 | - | 5 | 446 | ||
AY881621.1 | - | 5 | 446 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ykt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031692.2 | NP_113880.2 | 5 | 446 | ||
GenBank mRNA | AF033027.1 | - | 5 | 446 | ||
AY881621.1 | - | 5 | 446 |