siRNA Details
| Gene Symbol: Htr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024365.1 | NP_077341.1 | 1 | 500 | ||
| GenBank mRNA | L03202.1 | - | 1 | 619 | ||
| L19656.1 | - | 1 | 500 | |||
| L41146.1 | - | 1 | 182 | |||
| S62043.1 | - | 1 | 500 | |||
More Information
Set Membership: |
|

Htr6
Genomic Map