siRNA Details
| Gene Symbol: Cd200r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023953.1 | NP_076443.1 | 2, 3 | 118 | ||
| XM_006248347.2 | XP_006248409.1 | 2, 3 | 431 | |||
| GenBank mRNA | AY583211.1 | - | 2, 3 | 118 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd200r1| Gene Symbol: Cd200r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023953.1 | NP_076443.1 | 2, 3 | 118 | ||
| XM_006248347.2 | XP_006248409.1 | 2, 3 | 431 | |||
| GenBank mRNA | AY583211.1 | - | 2, 3 | 118 | ||