siRNA Details
| Gene Symbol: Mrpl23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022529.1 | NP_071974.1 | 4 | 272 | ||
| GenBank mRNA | BC059936.1 | - | 4 | 278 | ||
| U62635.1 | - | 4 | 272 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl23| Gene Symbol: Mrpl23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022529.1 | NP_071974.1 | 4 | 272 | ||
| GenBank mRNA | BC059936.1 | - | 4 | 278 | ||
| U62635.1 | - | 4 | 272 | |||