siRNA Details
| Gene Symbol: Prdx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022540.1 | NP_071985.1 | 2 | 116 | ||
| GenBank mRNA | AF106944.1 | - | 2 | 116 | ||
| BC060567.1 | - | 2 | 117 | 6922338 | ||
| FQ212826.1 | - | 2 | 96 | |||
| FQ219655.1 | - | 2 | 113 | |||

Prdx3
Genomic Map