siRNA Details
| Gene Symbol: Rhob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022542.1 | NP_071987.1 | 1 | 530 | ||
| GenBank mRNA | M74295.1 | - | 1 | 530 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhob| Gene Symbol: Rhob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022542.1 | NP_071987.1 | 1 | 530 | ||
| GenBank mRNA | M74295.1 | - | 1 | 530 | ||