siRNA Details
| Gene Symbol: Dapk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022546.1 | NP_071991.1 | 7 | 805 | ||
| XM_006240991.1 | XP_006241053.1 | 7 | 936 | |||
| XM_006240993.2 | XP_006241055.1 | 7 | 1043 | |||
| XM_008765129.1 | XP_008763351.1 | 7 | 1269 | |||
| GenBank mRNA | AJ006971.1 | - | 7 | 805 | ||
| BC062076.1 | - | 7 | 806 | 5599435 | ||

Dapk3
Genomic Map