siRNA Details
| Gene Symbol: Masp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172043.1 | NP_742040.1 | 11 | 1448 | ||
| GenBank mRNA | AJ277747.1 | - | 10 | 1390 | ||
| BC128700.1 | - | 11 | 1449 | 7324989 | ||
| Y18564.1 | - | 10 | 1227 | |||
| Y18565.1 | - | 4 | 373 | |||
| Y18566.1 | - | 8 | 980 | |||
| Y18571.1 | - | 5 | 455 | |||
| Y18572.1 | - | 10 | 1285 | |||

Masp2
Genomic Map