siRNA Details
| Gene Symbol: Surf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172068.1 | NP_742065.1 | 5 | 493 | ||
| XM_006233864.1 | XP_006233926.1 | 5 | 504 | |||
| XM_006233865.1 | XP_006233927.1 | 4 | 473 | |||
| GenBank mRNA | AF182952.1 | - | 5 | 493 | ||
| Gene Symbol: LOC100912008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233726.2 | XP_006233788.1 | 5 | 504 | ||
| XM_006233727.2 | XP_006233789.1 | 473 | ||||
| XM_006233728.2 | XP_006233790.1 | 495 | ||||

Surf1
Genomic Map