siRNA Details
| Gene Symbol: Entpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172030.1 | NP_742027.1 | 3 | 251 | ||
| XM_006233644.2 | XP_006233706.1 | 3 | 351 | |||
| XM_008761606.1 | XP_008759828.1 | 3 | 351 | |||
| GenBank mRNA | AF276940.1 | - | 3 | 251 | ||
| BC086558.1 | - | 3 | 261 | 7302083 | ||
| Y11835.1 | - | 3 | 250 | |||

Entpd2
Genomic Map