siRNA Details
| Gene Symbol: Plekhb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172033.2 | NP_742030.1 | 3 | 292 | ||
| XM_006229787.2 | XP_006229849.1 | 4 | 530 | |||
| XM_006229788.2 | XP_006229850.1 | 3 | 691 | |||
| XM_006229789.2 | XP_006229851.1 | 4 | 601 | |||
| XM_006229791.2 | XP_006229853.1 | 3 | 1163 | |||
| XM_008759795.1 | XP_008758017.1 | 3 | 687 | |||
| GenBank mRNA | AF081582.1 | - | 3 | 292 | ||
| AF118562.1 | - | 3 | 262 | |||
| BC078704.1 | - | 3 | 322 | 7124581 | ||

Plekhb1
Genomic Map