siRNA Details
| Gene Symbol: Fmod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080698.1 | NP_542429.1 | 2 | 892 | ||
| GenBank mRNA | X82152.1 | - | 2 | 892 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fmod| Gene Symbol: Fmod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080698.1 | NP_542429.1 | 2 | 892 | ||
| GenBank mRNA | X82152.1 | - | 2 | 892 | ||