siRNA Details
| Gene Symbol: Eif2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172029.2 | NP_742026.1 | 8, 9 | 792 | ||
| GenBank mRNA | BC081709.1 | - | 8, 9 | 792 | 7135576 | |
| FQ217438.1 | - | 8, 9 | 830 | |||
| U05821.1 | - | 8, 9 | 811 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif2b1| Gene Symbol: Eif2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172029.2 | NP_742026.1 | 8, 9 | 792 | ||
| GenBank mRNA | BC081709.1 | - | 8, 9 | 792 | 7135576 | |
| FQ217438.1 | - | 8, 9 | 830 | |||
| U05821.1 | - | 8, 9 | 811 | |||