siRNA Details
| Gene Symbol: Tkt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022592.1 | NP_072114.1 | 8 | 925 | ||
| XM_006252661.1 | XP_006252723.1 | 7 | 957 | |||
| GenBank mRNA | FQ232736.1 | - | 3 | 264 | ||
| U09256.1 | - | 8 | 925 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tkt| Gene Symbol: Tkt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022592.1 | NP_072114.1 | 8 | 925 | ||
| XM_006252661.1 | XP_006252723.1 | 7 | 957 | |||
| GenBank mRNA | FQ232736.1 | - | 3 | 264 | ||
| U09256.1 | - | 8 | 925 | |||