siRNA Details
| Gene Symbol: Tkt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022592.1 | NP_072114.1 | 7, 8 | 859 | ||
| XM_006252661.1 | XP_006252723.1 | 6, 7 | 891 | |||
| GenBank mRNA | FQ232736.1 | - | 2, 3 | 198 | ||
| U09256.1 | - | 7, 8 | 859 | |||

Tkt
Genomic Map