siRNA Details
| Gene Symbol: Pdap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022595.1 | NP_072117.1 | 3, 4 | 234 | ||
| GenBank mRNA | U26541.1 | - | 3, 4 | 234 | ||
| U41744.1 | - | 3, 4 | 255 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdap1| Gene Symbol: Pdap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022595.1 | NP_072117.1 | 3, 4 | 234 | ||
| GenBank mRNA | U26541.1 | - | 3, 4 | 234 | ||
| U41744.1 | - | 3, 4 | 255 | |||