siRNA Details
| Gene Symbol: Pdap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022595.1 | NP_072117.1 | 2 | 98 | ||
| GenBank mRNA | BC088140.1 | - | 2 | 112 | 7382664 | |
| U26541.1 | - | 2 | 98 | |||
| U41744.1 | - | 2 | 119 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdap1| Gene Symbol: Pdap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022595.1 | NP_072117.1 | 2 | 98 | ||
| GenBank mRNA | BC088140.1 | - | 2 | 112 | 7382664 | |
| U26541.1 | - | 2 | 98 | |||
| U41744.1 | - | 2 | 119 | |||