siRNA Details
| Gene Symbol: Ctsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022597.2 | NP_072119.2 | 7 | 790 | ||
| GenBank mRNA | BC072490.1 | - | 7 | 753 | 7097817 | |
| FQ212977.1 | - | 3 | 301 | |||
| M11305.1 | - | 4 | 454 | |||
| X82396.1 | - | 7 | 719 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctsb| Gene Symbol: Ctsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022597.2 | NP_072119.2 | 7 | 790 | ||
| GenBank mRNA | BC072490.1 | - | 7 | 753 | 7097817 | |
| FQ212977.1 | - | 3 | 301 | |||
| M11305.1 | - | 4 | 454 | |||
| X82396.1 | - | 7 | 719 | |||