siRNA Details
| Gene Symbol: Cnbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022598.1 | NP_072120.1 | 3 | 326 | ||
| XM_006236854.2 | XP_006236916.1 | 3 | 321 | |||
| GenBank mRNA | AB158421.1 | - | 2 | 161 | ||
| AB158422.1 | - | 2 | 161 | |||
| AF242550.1 | - | 3 | 212 | |||
| BC062225.1 | - | 3 | 310 | 6889682 | ||
| D45254.1 | - | 3 | 326 | |||

Cnbp
Genomic Map