siRNA Details
| Gene Symbol: Synj2bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022599.2 | NP_072121.2 | 2 | 218 | ||
| GenBank mRNA | AF260258.1 | - | 2 | 218 | ||
| BC126067.1 | - | 2 | 206 | 7323715 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Synj2bp| Gene Symbol: Synj2bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022599.2 | NP_072121.2 | 2 | 218 | ||
| GenBank mRNA | AF260258.1 | - | 2 | 218 | ||
| BC126067.1 | - | 2 | 206 | 7323715 | ||