siRNA Details
| Gene Symbol: B3gat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022609.2 | NP_072131.1 | 1 | 759 | ||
| GenBank mRNA | AB010441.1 | - | 1 | 759 | ||
| AF106624.1 | - | 1 | 754 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B3gat2| Gene Symbol: B3gat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022609.2 | NP_072131.1 | 1 | 759 | ||
| GenBank mRNA | AB010441.1 | - | 1 | 759 | ||
| AF106624.1 | - | 1 | 754 | |||