siRNA Details
| Gene Symbol: Icos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022610.1 | NP_072132.1 | 2 | 408 | ||
| XM_006245037.2 | XP_006245099.1 | 4 | 660 | |||
| XM_006245038.2 | XP_006245100.1 | 4 | 657 | |||
| XM_008767136.1 | XP_008765358.1 | 4 | 675 | |||
| GenBank mRNA | AB023133.1 | - | 2 | 408 | ||
| AB023134.1 | - | 2 | 408 | |||

Icos
Genomic Map