siRNA Details
| Gene Symbol: Top1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022615.1 | NP_072137.1 | 18 | 1859 | ||
| GenBank mRNA | AF140782.1 | - | 18 | 1859 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Top1| Gene Symbol: Top1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022615.1 | NP_072137.1 | 18 | 1859 | ||
| GenBank mRNA | AF140782.1 | - | 18 | 1859 | ||