siRNA Details
| Gene Symbol: Sept7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113740.1 | NP_001107212.1 | 5 | 485 | ||
| NM_022616.2 | NP_072138.2 | 4 | 482 | |||
| GenBank mRNA | AF142759.1 | - | 4 | 449 | ||
| BC128738.1 | - | 5 | 383 | 7381331 | ||
| EV764168.1 | - | 5 | 457 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009859, NM_001205367, AK147986, AK088997, AK144127, and others.

Sept7
Genomic Map