siRNA Details
| Gene Symbol: Fzd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022623.2 | NP_072145.1 | 2 | 1364 | ||
| GenBank mRNA | AF183910.1 | - | 2 | 1395 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fzd4| Gene Symbol: Fzd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022623.2 | NP_072145.1 | 2 | 1364 | ||
| GenBank mRNA | AF183910.1 | - | 2 | 1395 | ||
Set Membership: |
|