siRNA Details
| Gene Symbol: Slc22a23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022624.1 | NP_072146.1 | 4 | 1682 | ||
| XM_006253845.2 | XP_006253907.1 | 4 | 962 | |||
| GenBank mRNA | BC158843.1 | - | 4 | 581 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc22a23| Gene Symbol: Slc22a23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022624.1 | NP_072146.1 | 4 | 1682 | ||
| XM_006253845.2 | XP_006253907.1 | 4 | 962 | |||
| GenBank mRNA | BC158843.1 | - | 4 | 581 | ||