siRNA Details
| Gene Symbol: Phka1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022626.1 | NP_072148.1 | 26 | 2910 | ||
| XM_006257101.2 | XP_006257163.1 | 26 | 3237 | |||
| XM_006257102.2 | XP_006257164.1 | 25 | 3060 | |||
| GenBank mRNA | AF197561.1 | - | 26 | 2910 | ||

Phka1
Genomic Map