siRNA Details
| Gene Symbol: Phka1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022626.1 | NP_072148.1 | 7, 8 | 728 | ||
| GenBank mRNA | AF197561.1 | - | 7, 8 | 728 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phka1| Gene Symbol: Phka1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022626.1 | NP_072148.1 | 7, 8 | 728 | ||
| GenBank mRNA | AF197561.1 | - | 7, 8 | 728 | ||