siRNA Details
| Gene Symbol: Bbox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022629.1 | NP_072151.1 | 5 | 659 | ||
| GenBank mRNA | AF160958.1 | - | 5 | 659 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bbox1| Gene Symbol: Bbox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022629.1 | NP_072151.1 | 5 | 659 | ||
| GenBank mRNA | AF160958.1 | - | 5 | 659 | ||