siRNA Details
| Gene Symbol: Bbox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022629.1 | NP_072151.1 | 6, 7 | 1009 | ||
| GenBank mRNA | AF160958.1 | - | 6, 7 | 1009 | ||
| FQ210746.1 | - | 1101 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bbox1| Gene Symbol: Bbox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022629.1 | NP_072151.1 | 6, 7 | 1009 | ||
| GenBank mRNA | AF160958.1 | - | 6, 7 | 1009 | ||
| FQ210746.1 | - | 1101 | ||||